Learn More
IKBKG Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812458S

Description
IKBKG Monoclonal antibody specifically detects IKBKG in Human samples. It is validated for Western BlotSpecifications
IKBKG | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BRP1 | |
Mouse | |
Affinity chromatography | |
RUO | |
84306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI2F2 | |
Unconjugated | |
PDCD2L | |
1110037D23Rik; AI848108; AI851264; AMCBX1; AW124339; FIP3; FIP-3; Fip3p; gikk; I-kappa-B kinase gamma; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; ikB kinase-associated protein 1; IKBKG; IKK[g]; IKK3; IKKAP1; IKKG; IKK-gamma; IMD33; incontinentia pigmenti; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma; inhibitor of kappaB kinase gamma; inhibitor of nuclear factor kappa-B kinase subunit gamma; IP; IP1; IP2; IPD2; mF; mFIP-3; Nemo; NF-kappa B essential modulator; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188; ZC2HC9 | |
Full length human recombinant protein of human PDCD2L produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.