Learn More
H 44 Boss Head Clamp
Use this clamp with IKA™ magnetic stirrers. IKA H 44 Boss Head Clamps feature a sturdy, cast aluminum design.
Brand: IKA 0002437700
473.69 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
- Cast aluminum cross sleeve
- For fastening Holding Rod (Cat. No. 14-259-87) to Support Rod (14-259-79)
Specifications
For fastening the Holding rod H 38 to the support rod H 16 V | |
Boss Head Clamp | |
Aluminum |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30