Learn More
Proteintech IHCeasy NFIX Ready-To-Use IHC Kit
KHC1741 is a ready-to-use IHC kit for staining of NFIX.
Brand: Proteintech KHC1741-50T-S
This item is not returnable.
View return policy
Description
KHC1741 is a ready-to-use IHC kit for staining of NFIX. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
50 Tests | |
Liquid |