Learn More
Proteintech IHCeasy GTF2E1 Ready-To-Use IHC Kit
KHC1604 is a ready-to-use IHC kit for staining of GTF2E1.
Brand: Proteintech KHC1604-50T
This item is not returnable.
View return policy
Description
KHC1604 is a ready-to-use IHC kit for staining of GTF2E1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
50 Tests | |
Liquid |