Learn More
Proteintech IHCeasy CK8 Ready-To-Use IHC Kit
A ready-to-use IHC kit for the staining of CK8 (Cytokeratin 8).
Brand:  Proteintech KHC0041-50T
Product Code. 17496743
  • 4550.83 NOK / 1 set
Explore more special offers
Description
Description
  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
Human, Rat, Mouse
Store at 2°C to 8°C
50 Reactions
FFPE Tissue
50 Tests