missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IER5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556287
This item is not returnable.
View return policy
Description
Immunogen sequence: RRNLEQPPSG GEDDDAEEME TGNVANLISI FGSSFSGLLR KSPGGGREEE EGEESGPEAA EPGQICCDKP V Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 77%.
This gene encodes a protein that is similar to other immediate early response proteins. In the mouse, a similar gene may play an important role in mediating the cellular response to mitogenic signals. Studies in rats found the expression of a similar gene to be increased after waking and sleep deprivation.
Specifications
IER5 | |
Polyclonal | |
Unconjugated | |
IER5 | |
Ier5; immediate early response 5; immediate early response gene 5 protein; immediately early response; MGC102760; PP4583; SBBI48; si:dz129i22.1; Unknown (protein for MGC:136422); wu:fa99a02; wu:fb04b03; wu:fb11f08; zgc:136422 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51278 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VY09 | |
IER5 | |
Recombinant protein corresponding to Human IER5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IER5 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Spot an opportunity for improvement?Share a Content Correction