Learn More
BINDER™ ICH Light Module with LQC
ICH-Q1B light module with light-dose control
94698.51 NOK - 97490.61 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
30238296
|
BINDER™
8012-2443 |
KB PRO 260, KB PRO 470, KBF 260, KBF 470, KBF PRO 260, KBF PRO 470 |
94698.51 NOK
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
30238201
|
BINDER™
8012-2444 |
KB PRO 720, KBF 720, KBF PRO 720 |
97490.61 NOK
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
Module consists of:
- VIS cassette
- UVA cassette
- Spherical VIA sensor
- Spherical UVA sensor
- Independent power supply
Specifications
Glass | |
Lighting | |
LQC |
Product Title
BINDER™ ICH Light Module with LQC