missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15364-1-AP-150UL
This item is not returnable.
View return policy
Description
Intercellular adhesion molecule-1 (ICAM-1, CD54) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated (LFA) antigens, a beta 2 leukocyte integrin. The normal function of human ICAM-1 is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen (LFA-1) or macrophage-1 antigen (Mac-1). It is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses.Specifications
ICAM-1 | |
Polyclonal | |
Unconjugated | |
ICAM1 | |
BB2, CD54, ICAM 1, ICAM1, P3.58 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3383 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05362 | |
ICAM1 | |
ICAM-1 Fusion Protein Ag8309 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |