Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 1-10μL
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
2130.00 NOK - 2895.00 NOK
Specifications
For Use With (Application) | Microscale Solid Phase Extraction |
---|---|
For Use With (Equipment) | Micropipettes |
Product Line | HyperSep |
Type | SPE Tips |
Volume (Metric) | 1 to 10 μL |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
10600184
|
Thermo Scientific™
60109-201 |
BioBasic C18, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10703487
|
Thermo Scientific™
60109-202 |
BioBasic C8, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10610184
|
Thermo Scientific™
60109-203 |
BioBasic C4, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10456113
|
Thermo Scientific™
60109-204 |
Hypercarb, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10098952
|
Thermo Scientific™
60109-205 |
C18 plus Hypercarb, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10223233
|
Thermo Scientific™
60109-206 |
HILIC, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10426754
|
Thermo Scientific™
60109-207 |
Trypsin, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10059002
|
Thermo Scientific™
60109-208 |
Titanium Dioxide, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
10556005
|
Thermo Scientific™
60109-217 |
Zirconium Dioxide, 1 to 10 μL Volume | N/A | N/A | N/A | |||||
Description
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
- Mass spectrometry
- Desalting
- Protein purification
- MALDI
- Electrophoresis
- HPCE
- HPLC
- CEC
Tips are available in 1-10μL capacity and 10-200μL capacity.
Specifications
Microscale Solid Phase Extraction | |
HyperSep | |
1 to 10 μL |
Micropipettes | |
SPE Tips |