Learn More
Thermo Scientific™ HyperSep™ Cyano Cartridges
Get optimized isolation of polar compounds from nonpolar matrices, with these medium polarity cyano SPE cartridges.
2490.00 NOK - 4730.00 NOK
Specifications
For Use With (Application) | Retention of Polar Compounds From Nonpolar Matrices |
---|---|
For Use With (Equipment) | Thermo Scientific HyperSep-96 System |
Particle Size | 40 to 60 ÎĽm |
Product Line | HyperSep |
Sorbent | HyperSep Cyano |
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | |||||
10755564
|
Thermo Scientific™
60108-745 |
100 mg | 1 mL |
3655.00 NOK
Pack of 100 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
10556375
|
Thermo Scientific™
60108-746 |
50 mg | 1 mL |
3200.00 NOK
Pack of 100 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
10121083
|
Thermo Scientific™
60108-747 |
200 mg | 3 mL |
2690.00 NOK
Pack of 50 |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10765564
|
Thermo Scientific™
60108-748 |
500 mg | 3 mL |
2490.00 NOK
Pack of 50 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
10364233
|
Thermo Scientific™
60108-750 |
1000 mg | 6 mL |
2745.00 NOK
Pack of 30 |
Estimated Shipment: 15-09-2025
Log in to see stock. |
|||||
10239794
|
Thermo Scientific™
60108-751 |
2000 mg | 15 mL |
2805.00 NOK
Pack of 20 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||||
10417134
|
Thermo Scientific™
60108-752 |
5000 mg | 25 mL |
4730.00 NOK
Pack of 20 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
10630816
|
Thermo Scientific™
60108-753 |
10 g | 75 mL | N/A | ||||||
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano cartridges are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices. Typical applications include:
- Retention of polar compounds from hexane and oils
- Reversed phase extraction of moderately polar compounds
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
Retention of Polar Compounds From Nonpolar Matrices | |
40 to 60 ÎĽm | |
HyperSep Cyano |
Thermo Scientific HyperSep-96 System | |
HyperSep | |
Solid Phase Extraction Cartridge |