Learn More
Ansell™ HyFlex™ INTERCEPT™ Cut Resistant Medium Duty Industrial Sleeves w/o Thumbslot Product Code.: 16835410
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
Ansell™ HyFlex™ INTERCEPT™ Cut Resistant Medium Duty Industrial Sleeves w/o Thumbslot
GREENER_CHOICE
Product Code. 16835410
30.4 cm, Wide
Click to view available options
Length (Metric):
30.4 cm
40.6 cm
45.7 cm
Size:
Narrow
Wide
Length (English):
16 in.
18 in.
Product Code. 16835410
Brand: Ansell™ 11250120W
Estimated Shipment: 08-07-2025
Made of black yarn and are silicone and latex free for friendly paint and finish process
  • Seamless design
  • Red and black over edge is to indicate wide (black) and narrow (red) for easy laundry sorting purpose
  • Special knitted bicep cuffs keeps sleeve in place
  • Available in different lengths and widths (narrow or wide) for optimal fit and comfort to fit every worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Useful in automotive and transport, aerospace, machinery and equipment, metal fabrication, white goods industries
Applications:
  • Adjusting panels parts components
  • Adjusting systems and screwing
  • Body Surface checking, marking and touching up
  • Glass repair and replacement
  • Handling metal sheets and panels
  • Handling parts with sharp, rough edges
  • Handling structural and body components
  • Inspection, selecting, checking parts
  • Press shop operations
  • Press shop operations
  • Seizing structural and body parts
Compliance
Meets ANSI A3 (cut level) and EN 388-2542 standards.
Greener Choice Claims
  • Environmental benefits include:
    • Free-of a substance that causes environmental harm
    • Recyclable
TRUSTED_SUSTAINABILITY
Specifications
Size Wide
Product Type Cut-resistant Sleeve
Material Ansell INTERCEPT™ Technology
Color Dark Gray
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Seam Type Seamless
Latex Free Yes
Length (Metric) 30.4 cm
Product Title
Ansell™ HyFlex™ INTERCEPT™ Cut Resistant Medium Duty Industrial Sleeves w/o Thumbslot >