Learn More
Brannan™ Hydrometer specific gravity shot loaded medium adjustment for surface tension 1.300 to 1.350 x 0.001 S.G. 270 mm Brannan
Slot loaded density hydrometer for a variety of surface tension ranges.
Brand:  Brannan™ 10/930/0
Product Code. 12871468
  • 187.00 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Specifications
Specific Gravity Hydrometer
270 mm
Soda Glass
0.001 g/mL
1.30 to 1.35 g/mL
Steel Shot
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brannan™ Hydrometer specific gravity shot loaded medium adjustment for surface tension 1.300 to 1.350 x 0.001 S.G. 270 mm Brannan >