missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Hydrometer Only, for Battery Hydrometer Syphon Set
Designed for various industrial applications, including fleet maintenance, automotive services, solar-powered system maintenance, and general industrial uses, the Battery Hydrometer is utilized to measure battery acid levels accurately.
Brand: VEE GEE 6605-5H
This item is not returnable.
View return policy
Description
- Specific Gravity Range: 1.150 to 1.300
- Subdivisions: 0.005
- Approx. Length: 220 mm
- Calibrated for use at 25°C/77°F
Specifications
Hydrometer | |
22 cm | |
25°C (77°F) |
0.005 | |
1.150 to 1.300 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction