missing translation for 'onlineSavingsMsg'
Learn More
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals Product Code.: 11371689

Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals

Product Code. 11371689
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11371689

Brand: Thermo Scientific Alfa Aesar 012325.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27988-77-8
MDL Number MFCD00149903
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Packaging Glass bottle
Molecular Formula AuCl4H
Quantity 25 g
UN Number UN3260
Sensitivity Hygroscopic; Light sensitive
Synonym Tetrachloroauric(III) acid,Chloroauric acid
Solubility Information Soluble in water. Insoluble in ether.
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
Molecular Weight (g/mol) 339.77
Formula Weight 339.79 (anhydrous)
Percent Purity 99.9%
Physical Form Crystalline
Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP05L047 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number012325.06,012325.03,012325.14
Lot Number10253613 Certificate TypeCertificate of Analysis Date08/01/2025 Catalog Number012325.06,012325.03,012325.14
Lot NumberY25K054 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number012325.06,012325.03,012325.14
Lot NumberY01K027 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number012325.06,012325.03,012325.14
Lot Number10252562 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number012325.06,012325.03,012325.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.