missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen perOxide Solution, purum p.a., ≥35% (RT), Honeywell™

Purum p.a.,≥35% (RT)

450.08 NOK - 2474.99 NOK

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15677540
View Documents
Honeywell Research Chemicals
95299-500ML
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15657540
View Documents
Honeywell Research Chemicals
95299-1L
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15667540
View Documents
Honeywell Research Chemicals
95299-2.5L
2.5 L
2474.99 NOK
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338 + P310

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrogen perOxide Solution, purum p.a., ≥35% (RT), Honeywell™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.