missing translation for 'onlineSavingsMsg'
Learn More
Hydrogen Peroxide, 30% w/w, ACS Reagent Grade, Scharlab Product Code.: 11332635
compliance-icons compliance-icons

Hydrogen Peroxide, 30% w/w, ACS Reagent Grade, Scharlab

Product Code. 11332635
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11332635

Brand: Scharlab HI01361000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Used in analytical chemistry, as an oxidizing agent, as a bleaching agent and for pharmaceutical use. Scharlab Hydrogen Peroxide, 30% w/w, ACS Reagent Grade has a volume strength of 110.

  • ACS Reagent Grade
  • Synonyms: Hydrogen dioxide, Hydroperoxide
  • Molecular Formula: H2O2
  • Molar Mass: 34.01g/mol
  • CAS: 7722-84-1
  • EIN ECS No.: 231-765-0
  • Vapor Pressure: ∼18hPa (at 20°C)
  • Residue on Evaporation: max 0.002%

Specifications

Boiling Point 107°C
Melting Point -26°C
CAS 7722-84-1
Physical Form Solution
Quantity 1 L
Molecular Formula H2O2
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name hydrogen peroxide
Molecular Weight (g/mol) 34.014
PubChem CID 784
ChEBI CHEBI:16240
Formula Weight 34.01g/mol
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) 30.0–32% assay (permanganometric)
Density 1.11g/cm3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide, solution 30% w/w (110 vol), reagent grade, ACS, ISO
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrogen Peroxide, 30% w/w, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.