missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Appearance | Colorless |
pH | <1 |
Composition | Hydrochloric Acid 32% v.p. |
Concentration | 32+% HCl |
Density | 1.16g/mL |
Heavy Metals (as Pb) | <0.0001% |
Quantity | 1 L |
Physical Form | Liquid |
Solubility Information | Soluble in Water |
Percent Purity | >32% |
Safety and Handling


Product Identifier
- Hydrochloric acid 32% v.p.
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrochloric acid 32%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrochloric Acid, >32% Assay, Chem Lab™ >
Spot an opportunity for improvement?Share a Content Correction