Learn More
Hydrochloric Acid, 0.1M, Chem Lab™
484.00 NOK - 574.00 NOK
Specifications
Appearance | Colorless |
---|---|
pH | <2 |
Composition | Hydrochloric Acid 0.1mol/L Propanol-2 |
Concentration | 3.646g HCl/l C3H8O = 0.1 N (±0.0002/20°C) |
Density | 0.79g/mL |
Odor | 2-Propanol |
Physical Form | Liquid |
Solubility Information | Soluble in Water |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11933094
|
Chem Lab
CL05.0320.1000 |
1 L |
484.00 NOK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11953113
|
Chem Lab
CL05.0307.5500 |
5 L |
574.00 NOK
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Colorless | |
Hydrochloric Acid 0.1mol/L Propanol-2 | |
0.79g/mL | |
Liquid |
<2 | |
3.646g HCl/l C3H8O = 0.1 N (±0.0002/20°C) | |
2-Propanol | |
Soluble in Water |
Safety and Handling
Recommended Storage : Closed in a well ventilated place. Away from sources of ignition and heat.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrochloric acid 0.1 mol/l in Propanol-2
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H290-May be corrosive to metals.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid
Product Title
Hydrochloric Acid, 0.1M, Chem Lab™