missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYAL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | HYAL3 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16859124
|
Proteintech
19625-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16849124
|
Proteintech
19625-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the hyaluronidase family. Hyaluronidases are endoglycosidase enzymes that degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. The regulated turnover of hyaluro plays a critical role in many biological processes including cell proliferation, migration and differentiation. The encoded protein may also play an important role in sperm function. This gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression, and the expression of specific transcript variants may be indicative of tumor status. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and some isoforms may lack hyaluronidase activity. This gene overlaps and is on the same strand as N-acetyltransferase 6 (GCN5-related), and some transcripts of each gene share a portion of the first exon.Specifications
HYAL3 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43820, Q76HM9, Q8VEI3 | |
109685, 300993, 8372 | |
HYAL3 Fusion Protein Ag8611 | |
Primary | |
-20°C | |
HYAL3 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Hyal 3, HYAL3, Hyaluronidase 3, hyaluronoglucosaminidase 3, LUCA 3, LUCA14, LUCA3, Lung carcinoma protein 3, Minna14 | |
HYAL3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |