Learn More
R&D Systems™ Recombinant Human TSHR Fc Chimera Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 8950-TR-050
Encompass
Product Code. 15780324
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Recombinant Human TSH R Fc Chimera Protein is derived from CHO. The Recombinant Human TSH R Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
7253
TSHR
Chinese Hamster Ovary cell line,CHO-derived human TSH R protein, (N-terminus) Human TSH R (Met22-Gly413) Accession &Num; P16473 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
<0.10 EU per 1μg of the protein by the LAL method.
TSH R
Measured by its ability to inhibit rhTSH-induced cAMP accumulation in HEK293 human embryonic kidney cells transfected with human TSH R. The ED50 for this effect is typically 2-12μg/mL.
Yes
Theoretical M.W.: 71kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
CHNG1, LGR3, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH R, TSH-R
TSHR
Unconjugated
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human TSHR Fc Chimera Protein, CF