Learn More
Invitrogen™ Human TMEM70 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1029
This item is not returnable.
View return policy
Description
TMEM70 Synthetic Peptide, PEP-1029, from Invitrogen.
This peptide corresponds to 14 amino acids near the center of human TMEM70.PEP-1029 can be used as a blocking peptide with polyclonal antibody PA5-20915.Specifications
54968 | |
purified | |
Human TMEM70 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9BUB7 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TMEM70 | |
0.02% sodium azide | |
50 μg | |
PBS |