Learn More
R&D Systems™ Human TMED1 (NP_006849) VersaClone cDNA
R&D Systems™ Human TMED1 (NP_006849) VersaClone cDNA belongs to the TMED (transmembrane emp24 domain-containing) protein family.
Brand:  R&D Systems™ RDC2243
Product Code. 18758129
  • 3335.00 NOK / 10µg
Explore more special offers
Description
Description
The Human TMED1 (NP_006849) VersaClone cDNA clone contains the complete ORF for TMED1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TMED1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
Specifications
hTMED1
10 μg at 0.2 μg/μL
697bp
11018
Human
NP_006849
cDNA is provided in 10 mM Tris-Cl
8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.