missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TMCO6 Full-length ORF (AAH09618.1, 1 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH09618.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55374 |
Molecular Weight (g/mol) | 54.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181473
|
Abnova™
H00055374-P01.10ug |
10 ug |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16191473
|
Abnova™
H00055374-P01.25ug |
25 ug |
6245.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence: MLQMLQPGPKLNPGVAVEFAWCLHYIICSQVSNPLLIGHGALSTLGLLLLDLAGAVQKTEDAGLELLACPVLRCLSNLLTEAAVETVGGQMQLRDERVVAALFILLQFFFQKQPSLLPEGLWLLNNLTANSPSFCTSLLSLDLIEPLLQLLPVSNVVSVMVLTVLCNVAEKGPAYCQRLWPGPLLPALLHTLAFSDTEVVGQSLELLHLLFLYQPEAVQVFLQQSGLQALERHQEEAQLQDRVYALQQTALQGSpecifications
AAH09618.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLQMLQPGPKLNPGVAVEFAWCLHYIICSQVSNPLLIGHGALSTLGLLLLDLAGAVQKTEDAGLELLACPVLRCLSNLLTEAAVETVGGQMQLRDERVVAALFILLQFFFQKQPSLLPEGLWLLNNLTANSPSFCTSLLSLDLIEPLLQLLPVSNVVSVMVLTVLCNVAEKGPAYCQRLWPGPLLPALLHTLAFSDTEVVGQSLELLHLLFLYQPEAVQVFLQQSGLQALERHQEEAQLQDRVYALQQTALQG | |
RUO | |
TMCO6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55374 | |
TMCO6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ39769/PRO1580 | |
TMCO6 | |
Recombinant | |
wheat germ expression system |