Learn More
Abnova™ Human TAP2 Full-length ORF (AAH02751, 1 a.a. - 686 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006891-P01.25ug
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. This gene is located 7 kb telomeric to gene family member ABCB2. The protein encoded by this gene is involved in antigen presentation. This protein forms a heterodimer with ABCB2 in order to transport peptides from the cytoplasm to the endoplasmic reticulum. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease. Alternative splicing of this gene produces two products which differ in peptide selectivity and level of restoration of surface expression of MHC class I molecules. [provided by RefSeq]
Sequence: MRLPDLRPWTSLLLVDAALLWLLQGPLGTLLPQGLPGLWLEGTLRLGGLWGLLKLRGLLGFVGTLLLPLCLATPLTVSLRALVAGASRAPPARVASAPWSWLLVGYGAAGLSWSLWAVLSPPGAQEKEQDQVNNKVLMWRLLKLSRPDLPLLVAAFFFLVLAVLGETLIPHYSGRVIDILGGDFDPHAFASAIFFMCLFSFGSSLSAGCRGGCFTYTMSRINLRIREQLFSSLLRQDLGFFQETKTGELNSRLSSDTTLMSNWLPLNANVLLRSLVKVVGLYGFMLSISPRLTLLSLLHMPFTIAAEKVYNTRHQEVLREIQDAVARAGQVVREAVGGLQTVRSFGAEEHEVCRYKEALEQCRQLYWRRDLERALYLLVRRVLHLGVQMLMLSCGLQQMQDGELTQGSLLSFMIYQESVGSYVQTLVYIYGDMLSNVGAAEKVFSYMDRQPNLPSPGTLAPTTLQGVVKFQDVSFAYPNRPDRPVLKGLTFTLRPGEVTALVGPNGSGKSTVAALLQNLYQPTGGQVLLDEKPISQYEHCYLHSQVVSVGQEPVLFSGSVRNNIAYGLQSCEDDKVMAAAQAAHADDFIQEMEHGIYTDVGEKGSQLAAGQKQRLAIARALVRDPRVLILDEATSALDVQCEQALQDWNSRGDRTVLVIAHRLQTVQRAHQILVLQEGKLQKLAQLSpecifications
AAH02751 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
101.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC18/ABCB3/APT2/D6S217E/PSF2/RING11 | |
TAP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6891 | |
TAP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRLPDLRPWTSLLLVDAALLWLLQGPLGTLLPQGLPGLWLEGTLRLGGLWGLLKLRGLLGFVGTLLLPLCLATPLTVSLRALVAGASRAPPARVASAPWSWLLVGYGAAGLSWSLWAVLSPPGAQEKEQDQVNNKVLMWRLLKLSRPDLPLLVAAFFFLVLAVLGETLIPHYSGRVIDILGGDFDPHAFASAIFFMCLFSFGSSLSAGCRGGCFTYTMSRINLRIREQLFSSLLRQDLGFFQETKTGELNSRLSSDTTLMSNWLPLNANVLLRSLVKVVGLYGFMLSISPRLTLLSLLHMPFTIAAEKVYNTRHQEVLREIQDAVARAGQVVREAVGGLQTVRSFGAEEHEVCRYKEALEQCRQLYWRRDLERALYLLVRRVLHLGVQMLMLSCGLQQMQDGELTQGSLLSFMIYQESVGSYVQTLVYIYGDMLSNVGAAEKVFSYMDRQPNLPSPGTLAPTTLQGVVKFQDVSFAYPNRPDRPVLKGLTFTLRPGEVTALVGPNGSGKSTVAALLQNLYQPTGGQVLLDEKPISQYEHCYLHSQVVSVGQEPVLFSGSVRNNIAYGLQSCEDDKVMAAAQAAHADDFIQEMEHGIYTDVGEKGSQLAAGQKQRLAIARALVRDPRVLILDEATSALDVQCEQALQDWNSRGDRTVLVIAHRLQTVQRAHQILVLQEGKLQKLAQL | |
RUO | |
TAP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |