missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human STK17A Full-length ORF (AAH47696.1, 1 a.a. - 414 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009263-P01.25ug
This item is not returnable.
View return policy
Description
This gene is a member of the DAP kinase-related apoptosis-inducing protein kinase family and encodes an autophosphorylated nuclear protein with a protein kinase domain. The protein has apoptosis-inducing activity. [provided by RefSeq]
Sequence: MIPLEKPGSGGSSPGATSGSGRAGRGLSGPCRPPPPPQARGLLTEIRAVVRTEPFQDGYSLCPGRELGRGKFAVVRKCIKKDSGKEFAAKFMRKRRKGQDCRMEIIHEIAVLELAQDNPWVINLHEVYETASEMILVLEYAAGGEIFDQCVADREEAFKEKDVQRLMRQILEGVHFLHTRDVVHLDLKPQNILLTSESPLGDIKIVDFGLSRILKNSEELREIMGTPEYVAPEILSYDPISMATDMWSIGVLTYVMLTGISPFLGNDKQETFLNISQMNLSYSEEEFDVLSESAVDFIRTLLVKKPEDRATAEECLKHPWLTQSSIQEPSFRMEKALEEANALQEGHSVPEINSDTDKSETEESIVTEELIVVTSYTLGQCRQSEKEKMEQKAISKRFKFEEPLLQEIPGEFIYSpecifications
AAH47696.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRAK1 | |
STK17A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9263 | |
STK17A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIPLEKPGSGGSSPGATSGSGRAGRGLSGPCRPPPPPQARGLLTEIRAVVRTEPFQDGYSLCPGRELGRGKFAVVRKCIKKDSGKEFAAKFMRKRRKGQDCRMEIIHEIAVLELAQDNPWVINLHEVYETASEMILVLEYAAGGEIFDQCVADREEAFKEKDVQRLMRQILEGVHFLHTRDVVHLDLKPQNILLTSESPLGDIKIVDFGLSRILKNSEELREIMGTPEYVAPEILSYDPISMATDMWSIGVLTYVMLTGISPFLGNDKQETFLNISQMNLSYSEEEFDVLSESAVDFIRTLLVKKPEDRATAEECLKHPWLTQSSIQEPSFRMEKALEEANALQEGHSVPEINSDTDKSETEESIVTEELIVVTSYTLGQCRQSEKEKMEQKAISKRFKFEEPLLQEIPGEFIY | |
RUO | |
STK17A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |