Learn More
Invitrogen™ Human SKA1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0931
This item is not returnable.
View return policy
Description
SKA1 Synthetic Peptide, PEP-0931, from Invitrogen.
This peptide corresponds to 13 amino acids near the carboxy terminus of human SKA1.PEP-0931 can be used as a blocking peptide with polyclonal antibody PA5-20817.Specifications
220134 | |
purified | |
Human SKA1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q96BD8 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
SKA1 | |
0.02% sodium azide | |
50 μg | |
PBS |