Learn More
Invitrogen™ Human SKA1 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP0931
Product Code. 15389170
  • 1040.00 NOK / 50µg
Explore more special offers
Description
Description
SKA1 Synthetic Peptide, PEP-0931, from Invitrogen.
This peptide corresponds to 13 amino acids near the carboxy terminus of human SKA1.PEP-0931 can be used as a blocking peptide with polyclonal antibody PA5-20817.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
220134
purified
Human SKA1
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q96BD8
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
SKA1
0.02% sodium azide
50 μg
PBS