missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human POGK Full-length ORF (NP_060012.3, 1 a.a. - 609 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057645-P01.10ug
This item is not returnable.
View return policy
Description
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human. [provided by RefSeq]
Sequence: MESTAYPLNLSLKEEEEEEEIQSRELEDGPADMQKVRICSEGGWVPALFDEVAIYFSDEEWEVLTEQQKALYREVMRMNYETVLSLEFPFPKPDMITRLEGEEESQNSDEWQLQGGTSAENEESDVKPPDWPNPMNATSQFPQPQHFDSFGLRLPRDITELPEWSEGYPFYMAMGFPGYDLSADDIAGKFQFSRGMRRSYDAGFKLMVVEYAESTNNCQAAKQFGVLEKNVRDWRKVKPQLQNAHAMRRAFRGPKNGRFALVDQRVAEYVRYMQAKGDPITREAMQLKALEIAQEMNIPEKGFKASLGWCRRMMRRYDLSLRHKVPVPQHLPEDLTEKLVTYQRSVLALRRAHDYEVAQMGNADETPICLEVPSRVTVDNQGEKPVLVKTPGREKLKITAMLGVLADGRKLPPYIILRGTYIPPGKFPSGMEIRCHRYGWMTEDLMQDWLEVVWRRRTGAVPKQRGMLILNGFRGHATDSVKNSMESMNTDMVIIPGGLTSQLQVLDVVVYKPLNDSVRAQYSNWLLAGNLALSPTGNAKKPPLGLFLEWVMVAWNSISSESIVQGFKKCHISSNLEEEDDVLWEIESELPGGGEPPKDCDTESMAESNSpecifications
NP_060012.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BASS2/KIAA1513/KIAA15131/LST003 | |
POGK | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
57645 | |
POGK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESTAYPLNLSLKEEEEEEEIQSRELEDGPADMQKVRICSEGGWVPALFDEVAIYFSDEEWEVLTEQQKALYREVMRMNYETVLSLEFPFPKPDMITRLEGEEESQNSDEWQLQGGTSAENEESDVKPPDWPNPMNATSQFPQPQHFDSFGLRLPRDITELPEWSEGYPFYMAMGFPGYDLSADDIAGKFQFSRGMRRSYDAGFKLMVVEYAESTNNCQAAKQFGVLEKNVRDWRKVKPQLQNAHAMRRAFRGPKNGRFALVDQRVAEYVRYMQAKGDPITREAMQLKALEIAQEMNIPEKGFKASLGWCRRMMRRYDLSLRHKVPVPQHLPEDLTEKLVTYQRSVLALRRAHDYEVAQMGNADETPICLEVPSRVTVDNQGEKPVLVKTPGREKLKITAMLGVLADGRKLPPYIILRGTYIPPGKFPSGMEIRCHRYGWMTEDLMQDWLEVVWRRRTGAVPKQRGMLILNGFRGHATDSVKNSMESMNTDMVIIPGGLTSQLQVLDVVVYKPLNDSVRAQYSNWLLAGNLALSPTGNAKKPPLGLFLEWVMVAWNSISSESIVQGFKKCHISSNLEEEDDVLWEIESELPGGGEPPKDCDTESMAESN | |
RUO | |
POGK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |