Learn More
Invitrogen™ Human NOD2 Synthetic Peptide
Product Code. 15246519
50 μg, 50µg
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Synthetic Peptide
NOD2 Synthetic Peptide, PEP-0306, from Invitrogen.
NOD2 is a member of the apoptosis regulating protein family that includes caspase recruitment-domains, as well as Apaf-1 and NOD1. It contains two N-terminal CARDs, a nucleotide binding domain (NBD), and multiple C-terminal leucine-rich repeats (LRRs). NOD2 is expressed in monocytes (whereas NOD1 is expressed in multiple tissues). NOD2 plays a role in regulating NF-kappaB, acts as an intracellular receptor for bacterial lipopolysaccharides, and contributes to inflammatory bowel disease (IBD) and Crohn's disease.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 64127
Species Human
Purification Method purified
Accession Number Q9HC29
Common Name Human NOD2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol NOD2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Product Title
Invitrogen™ Human NOD2 Synthetic Peptide >