missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NME7 Partial ORF (NP_004547, 277 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029922-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: DRVNVEEFYEVYKGVVTEYHDMVTEMYSGPCVAMEIQQNNATKTFREFCGPADPEIARHLRPGTLRAIFGKTKIQNAVHCTDLPEDGLLEVQYFFKILSpecifications
NP_004547 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DRVNVEEFYEVYKGVVTEYHDMVTEMYSGPCVAMEIQQNNATKTFREFCGPADPEIARHLRPGTLRAIFGKTKIQNAVHCTDLPEDGLLEVQYFFKIL | |
RUO | |
NME7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29922 | |
NME7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ37194/nm23-H7 | |
NME7 | |
Recombinant | |
wheat germ expression system |