Specifications
Physical Form | Frozen |
Quantity | 0.5 mL |
Purity | Research Grade |
Host Species | Human |
Product Type | Human Vesicle |
Content And Storage | Store at -80°C. |
Format | Vial |
Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ Human MRP1 Vesicles >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu