missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human/Mouse/Porcine MYO7A (aa 16-31) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP045
This item is not returnable.
View return policy
Description
Mouse, Pig, Human Myosin Vlla (residues 16-31) Synthetic Peptide, PEP-045, from Invitrogen.
PEP-045 is a 16 amino acid synthetic peptide whose sequence corresponds to a sequence found in the N-terminal region, residues 16-31, of murine, porcine, and human myosin VIIa. The sequence of this peptide is (amino to carboxy terminus): S16-G-Q-E-F-D-V-P-I-G-A-V-V-K-L-C31This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and myosin VIIa, catalog # PA1-936. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
17921, 397373, 4647 | |
purified | |
Human/Mouse/Porcine MYO7A (aa 16-31) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human, Mouse, Porcine | |
P97479, Q13402, Q28970 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
MYO7A | |
No preservative | |
50 μg | |
S16 - G - Q - E - F - D - V - P - I - G - A - V - V - K - L - C31 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human/Mouse/Porcine MYO7A (aa 16-31) Synthetic Peptide > 50μg
Spot an opportunity for improvement?Share a Content Correction