Learn More
R&D Systems™ Human MCAM/CD146 (NP_006491) VersaClone cDNA
Melanoma Cell Adhesion Molecule.
Brand:  R&D Systems™ RDC1843
Product Code. 18732799
  • 7745.00 NOK / 10µg
Explore more special offers
Description
Description
The Human MCAM/CD146 (NP_006491) VersaClone cDNA clone contains the complete ORF for MCAM/CD146, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MCAM/CD146 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
cDNA
NP_006491
cDNA is provided in 10mM Tris-Cl, pH 8.5
4162
Human
hMCAM/CD146
10 μg at 0.2 μg/μL
1954bp
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in Humans.