missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LRRFIP1 Partial ORF (NP_004726, 675 a.a. - 784 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009208-Q01.10ug
This item is not returnable.
View return policy
Description
Sequence: GVAKDNAKIDGATQSSPAEPKSEDADRCTLPEHESPSQDISDACEAESTERCEMSEHPSQTVRKALDSNSLENDDLSAPGREPGHFNPESREDTRGGNEKGKSKEDCTMSSpecifications
NP_004726 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GVAKDNAKIDGATQSSPAEPKSEDADRCTLPEHESPSQDISDACEAESTERCEMSEHPSQTVRKALDSNSLENDDLSAPGREPGHFNPESREDTRGGNEKGKSKEDCTMS | |
RUO | |
LRRFIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9208 | |
LRRFIP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLAP-1/FLIIAP1/GCF-2/GCF2/HUFI-1/MGC10947/MGC119738/MGC119739/TRIP | |
LRRFIP1 | |
Recombinant | |
wheat germ expression system |