missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ITFG1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8900-SP
This item is not returnable.
View return policy
Description
ITFG1 Monoclonal specifically detects ITFG1 in Human samples. It is validated for Western Blot.
Specifications
ITFG1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CDA08, integrin alpha FG-GAP repeat containing 1,2310047C21Rik, Integrin-alpha FG-GAP repeat-containing protein 1, Protein TIP, T-cell immunomodulatory protein, TIP, TIPT cell immunomodulatory protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
81533 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
936234 | |
Western Blot 2 ug/mL | |
Q8TB96 | |
ITFG1 | |
Human embryonic kidney cell line HEK293-derived human ITFG1 Met1-Ile566 Accession # Q8TB96 | |
25 μg | |
Primary | |
Detects human ITFG1 in direct ELISAs. | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human ITFG1 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction