missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HHLA3 Partial ORF (NP_009002, 1 a.a. - 53 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011147-Q01.10ug

 View more versions of this product

Product Code. 16193776

  • 4115.00 NOK / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MFGACYKQPLKPSGSEPPAEECRMTPRHAGCDVTEMQRILSQPTFTEHLLRAV
Specifications

Specifications

NP_009002
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.57kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MFGACYKQPLKPSGSEPPAEECRMTPRHAGCDVTEMQRILSQPTFTEHLLRAV
RUO
HHLA3
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
11147
HHLA3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HHLA3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers