Learn More
Novus Biologicals™ Human Heart Atrium Whole Tissue Lysate (Adult Whole Arrhythmia)
Brand:  Novus Biologicals™ NB820-59310
Product Code. 18241455
  • 6380.00 NOK / 1mg
Explore more special offers
Description
Description
The Human Heart Atrium Whole Tissue Lysate (Adult Whole Arrhythmia) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Arrhythmia, infarct: Heart Atrium, right
Arrhythmia
Human
Human Heart Atrium Whole Tissue Lysate (Adult Arrhythmia)
Adult
Heart
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native