missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Galectin-9 ELISA Kit
ELISA
5880.00 NOK - 28720.00 NOK
Specifications
Accession Number | 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; Ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; Tumor antigen HOM-HD-21; UAT; Urate transporter/channel; urate transporter/channel protein |
---|---|
Assay Range | 36.86 to 9,000 pg/mL |
Assay Sensitivity | 36 pg/mL |
Conjugate | HRP |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17184959
|
Invitrogen™
EH206RBX5 |
5 x 96 Tests |
28720.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17134493
|
Invitrogen™
EH206RB |
96 Tests |
5880.00 NOK
Pack of 96 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human Galectin-9 quantitates human Galectin-9 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Galectin-9.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H and RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; Ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; Tumor antigen HOM-HD-21; UAT; Urate transporter/channel; urate transporter/channel protein | |
36 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Lgals9 | |
<10 | |
HRP | |
Plasma 50 ÎĽL, Serum 50 ÎĽL, Supernatant 50 ÎĽL | |
Galectin 9 | |
4 hrs 45 mins |
36.86 to 9,000 pg/mL | |
HRP | |
Plasma, Serum, Supernatant | |
3965 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human Galectin-9 ELISA Kit