Learn More
Invitrogen™ Human FKBP5 (aa 394-407) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP078
This item is not returnable.
View return policy
Description
Human FKBP51 (residues 394-407) Synthetic Peptide, PEP-078, from Invitrogen.
PEP-078 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 394-407 of human FKBP51 with a carboxy terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C (394) -Q-K-K-A-K-E-H-N-E-R-D-R-R (407)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human FKBP51, catalog # PA1-020. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
2289 | |
purified | |
Human FKBP5 (aa 394-407) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q13451 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
FKBP5 | |
No preservative | |
50 μg | |
C(394) -Q - K - K - A - K - E - H - N - E - R - D - R - R(407) |