missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FGF-17 ELISA Kit
Human FGF-17 quantitates human FGF-17 in serum, plasma, supernatant.
5430.00 NOK - 39970.00 NOK
Specifications
Accession Number | FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187 |
---|---|
Assay Range | 0.25 to 60 ng/mL |
Assay Sensitivity | 0.25 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17154939
|
Invitrogen™
EH186RBX5 |
5 x 96 Tests |
26530.00 NOK
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
17824090
|
Invitrogen™
EH186RBX10 |
10 x 96 Tests |
39970.00 NOK
Pack of 10 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
17104473
|
Invitrogen™
EH186RB |
96 Tests |
5430.00 NOK
Pack of 96 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
Human FGF-17 quantitates human FGF-17 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FGF-17.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.Specifications
FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187 | |
0.25 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
FGF17 | |
<10% | |
HRP | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
FGF17 | |
4 hr 45 min |
0.25 to 60 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O60258 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title