Learn More
Invitrogen™ Human FAM91A1 (aa 607-686) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP93961
Product Code. 30207473
  • 3015.00 NOK / 100µL
Explore more special offers
Description
Description
FAM91A1 (family with sequence similarity 91, member A1) is a 94 kDa phospho-protein that is not yet characterized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q658Y4
Blocking Assay, Control
157769
100 μL
RUO
FAM91A1
Human
LHGIGETVHVPFPFDETELQGEFTRVNMGVHKALQILRNRVDLQHLCGYVTMLNASSQLADRKLSDASDERGEPDLASGS
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
FAM91A1
-20° C, Avoid Freeze/Thaw Cycles
AV220772; BC033609; D15Ertd621e; DNA segment, Chr 15, ERATO Doi 621, expressed; Fam91a1; family with sequence similarity 91 member A1; family with sequence similarity 91, member A1; hypothetical protein FLJ23790; hypothetical protein LOC393413; Kiaa0493; mKIAA0493; Protein FAM91A1; si:dkey-5e12.2; skeletal muscle cells re-entry induced; SKIN; zgc:63609
Unconjugated
Recombinant
E. coli