Learn More
Invitrogen™ Human FAM91A1 (aa 607-686) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP93961
This item is not returnable.
View return policy
Description
FAM91A1 (family with sequence similarity 91, member A1) is a 94 kDa phospho-protein that is not yet characterized.Specifications
Q658Y4 | |
Blocking Assay, Control | |
157769 | |
100 μL | |
RUO | |
FAM91A1 | |
Human | |
LHGIGETVHVPFPFDETELQGEFTRVNMGVHKALQILRNRVDLQHLCGYVTMLNASSQLADRKLSDASDERGEPDLASGS | |
Liquid |
≥5.0 mg/mL | |
1M urea, PBS with no preservative; pH 7.4 | |
FAM91A1 | |
-20° C, Avoid Freeze/Thaw Cycles | |
AV220772; BC033609; D15Ertd621e; DNA segment, Chr 15, ERATO Doi 621, expressed; Fam91a1; family with sequence similarity 91 member A1; family with sequence similarity 91, member A1; hypothetical protein FLJ23790; hypothetical protein LOC393413; Kiaa0493; mKIAA0493; Protein FAM91A1; si:dkey-5e12.2; skeletal muscle cells re-entry induced; SKIN; zgc:63609 | |
Unconjugated | |
Recombinant | |
E. coli |