missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DSCAM ELISA Kit
Human DSCAM quantitates human DSCAM in serum, plasma, supernatant.
Brand: Invitrogen™ EH163RB
This item is not returnable.
View return policy
Description
Human DSCAM quantitates human DSCAM in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSCAM.
This gene is a member of the immunoglobulin superfamily of cell adhesion molecules (Ig-CAMs), and is involved in human central and peripheral nervous system development. This gene is a candidate for Down syndrome and congenital heart disease (DSCHD). A gene encoding a similar Ig-CAM protein is located on chromosome 11. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Oct 2012].Specifications
4932410A21Rik; CHD2; CHD2-42; CHD2-52; Down syndrome cell adhesion molecule; Down syndrome cell adhesion molecule homolog; DS cell adhesion molecule; Dscam | |
1.4 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
DSCAM | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
1.4 to 300 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
O60469 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
DSCAM | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction