missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CXCL9 (MIG) Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87843
This item is not returnable.
View return policy
Description
MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.Specifications
Q07325 | |
Control, ELISA, Western Blot | |
4283 | |
Human CXCL9 (aa 23-125) | |
E. coli | |
RUO | |
BB139920; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; Cxcl9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; MIG; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; Protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9 | |
Human | |
Recombinant | |
>95% |
200 μg/mL | |
PBS with no preservative | |
11.7 kD | |
100 μg | |
-20°C | |
0.01 ng/ μg cytokine | |
CXCL9 | |
Unconjugated | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction