missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CXCL10 (IP-10) Recombinant Protein

E. coli Recombinant Protein

1282.00 NOK - 19540.00 NOK

Specifications

Accession Number P02778
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 3627
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17606635
View Documents Promotion Details
Invitrogen™
RP87836
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17616635
View Documents Promotion Details
Invitrogen™
RP87837
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17626635
View Documents Promotion Details
Invitrogen™
RP87838
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17636635
View Documents Promotion Details
Invitrogen™
RP87839
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
Specifications

Specifications

P02778
Control, ELISA, Western Blot
3627
Human CXCL10 (aa 22-98)
-20°C
0.01 ng/ μg cytokine
CXCL10
Unconjugated
Liquid
200 μg/mL
PBS with no preservative
8.7 kD
E. coli
RUO
10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C x C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; crg-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamm
Human
Recombinant
>95%
Videos
SDS
Documents

Documents

Product Certifications