Learn More
R&D Systems™ Human Complement Factor D Magnetic Luminex™ Performance Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems™ LOBM1824
Product Code. 18462490
  • 2015.00 NOK / 1 set
Explore more special offers
Specifications
1 Kit
1675
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Human
Adipsin, ADN, ADNcomplement factor D, AMBP-1, C3 convertase activator, CFD, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human Complement Factor D Magnetic Luminex™ Performance Assay > 1 Kit