Learn More
Novus Biologicals™ Human Colon Tissue MicroArray (Tumor)
The panel contains 5 to 8 different tissues on one slide.
Brand:  Novus Biologicals™ NBP2-78088
Product Code. 18601571
  • 5670.00 NOK / Pack of 5
Explore more special offers
Specifications
5 Slides
Human
Please bake slides at 60C for 30 minutes before use.