Learn More
ACROBiosystems Human CD3 epsilon & CD3 delta Heterodimer Protein, His Tag&Tag Free
MALS verified

4968.94 NOK - 50212.48 NOK
Specifications
Accession Number | NP_000724.1 (CD3E) and NP_000723.1 (CD3D) |
---|---|
Molecular Weight (g/mol) | 16.6 kDa (CD3E) and 13.4 kDa (CD3D) |
Source | Human CD3E&CD3D; Heterodimer Protein, His Tag&Tag; Free (CDD-H52W1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) and P04234-1 (CD3D)). |
Immunogen | Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17136211
![]() |
ACROBiosystems
CDD-H52W1-1mg |
1 mg |
50212.48 NOK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16668512
![]() |
ACROBiosystems
CDD-H52W1-50UG |
50 ÎĽg |
4968.94 NOK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free is produced by co-expression of CD3E and CD3D, has a calculated MW of 16.6 kDa (CD3E) and 13.4 kDa (CD3D). Subunit CD3E is fused with a polyhistidine tag at the C-terminus and subunit CD3D contains noSpecifications
NP_000724.1 (CD3E) and NP_000723.1 (CD3D) | |
Human CD3E&CD3D; Heterodimer Protein, His Tag&Tag; Free (CDD-H52W1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) and P04234-1 (CD3D)). | |
-20°C | |
CD3E and CD3D | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
16.6 kDa (CD3E) and 13.4 kDa (CD3D) | |
Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) | |
CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / ÎĽg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.