Learn More
R&D Systems™ Human CD25/IL-2R alpha Magnetic Luminex™ Performance Assay
Brand:  R&D Systems™ LBAM223
Product Code. 30002079
  • 2015.00 NOK / 1 set
Explore more special offers
Specifications
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Human
3559
1 Kit
CD25/IL-2R alpha
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human CD25/IL-2R alpha Magnetic Luminex™ Performance Assay > 1 Kit