Learn More
R&D Systems™ Human AMPK alpha 1 (NP_006242) VersaClone cDNA
R&D Systems™ Human AMPK alpha 1 (NP_006242) VersaClone cDNA belongs to the ser/thr protein kinase family.
Brand: R&D Systems™ RDC2339
This item is not returnable.
View return policy
Description
The Human AMPK alpha 1 (NP_006242) VersaClone cDNA clone contains the complete ORF for AMPK alpha 1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The AMPK alpha 1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
AMPK alpha 1 | |
10 μg at 0.2 μg/μL | |
1693bp | |
5562 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_006242 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5 '-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5 '-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, PRKAA1, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
Human |