missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565031
This item is not returnable.
View return policy
Description
Immunogen sequence: MEEPGAQCAPP PPAGSETWVP QANLSSAPSQ NCSAKDYIYQ DS Highest antigen sequence identity to the following orthologs - mouse 67%, rat 72%.
Serotonin (5-hydroxytryptamine, 5-HT), originally discovered as a serum factor plays important roles in regulating diverse biological processes in central and peripheral nervous systems, cardiovascular systems, and gastrointestinal systems. Molecular cloning studies have identified 5 receptors belonging to the 5-HT1 subtypes. These receptors are negatively coupled to adenylyl cyclase. Radioligand binding studies suggest the presence of 5-HT1B (formerly known as 5-HT1DbR) sites in substantia nigra, dorsal subiculum, globus pallidus, and superior collicum. In situ hybridization studies show abundance of 5-HT1BR mRNA in CA1 pyramidal cells of hippocampus, Purkinje cells of cerebellum, olfactory tubercle, subthalamic nucleus and somewhat lower levels in the caudate-putamen, cortical areas, thalamic and hypothalamic nuclei.
Specifications
HTR1B | |
Polyclonal | |
Unconjugated | |
HTR1B | |
5-HT<1B> receptor; 5ht1b; 5-HT1B; 5-HT-1B; 5HT1B Receptor; 5-HT1B receptor; 5-HT1B serotonin receptor; 5-HT1DB; 5-HT-1D-beta; 5-hydroxytryptamine (serotonin ) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; HTR1B; HTR1D2; HTR1DB; S12; serotonin (5-hydroxytryptamine (5HT)) receptor type 1B; serotonin 1D beta receptor; Serotonin receptor 1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3351 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P28222 | |
HTR1B | |
Recombinant Human HTR1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HTR1B Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction