missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP47 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Specifications
Antigen | HSP47 |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SERPINH1 is a member of the serpin superfamily of serine proteinase inhibitors, and its expression is induced by heat shock. The SERPINH1 protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to SERPINH1 have been found in patients with rheumatoid arthritis.Specifications
HSP47 | |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P19324, P50454 | |
12406, 871 | |
HSP47 Fusion Protein Ag1301 | |
Primary | |
-20°C | |
SERPINH1 |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
47 kDa heat shock protein, AsTP3, CBP1, CBP2, Collagen binding protein, Colligin, gp46, HSP47, PIG14, PPROM, RA A47, Serpin H1, SERPINH1, SERPINH2 | |
SERPINH1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |